The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042599.1 | NP_001036064.1 | 5 - 6 | 723 | 62 |
NM_005235.2 | NP_005226.1 | 5 - 6 | 723 | 62 | |
XM_005246376.2 | XP_005246433.1 | 5 - 6 | 1242 | 62 | |
XM_005246377.2 | XP_005246434.1 | 5 - 6 | 1242 | 62 | |
XM_006712364.2 | XP_006712427.1 | 5 - 6 | 1242 | 62 | |
XM_017003577.1 | XP_016859066.1 | 6 - 7 | 1320 | 62 | |
XM_017003578.1 | XP_016859067.1 | 6 - 7 | 1320 | 62 | |
XM_017003579.1 | XP_016859068.1 | 6 - 7 | 1320 | 62 | |
XM_017003580.1 | XP_016859069.1 | 6 - 7 | 1320 | 62 | |
XM_017003581.1 | XP_016859070.1 | 6 - 7 | 1320 | 62 | |
GenBank mRNA | AK308606.1 | - | 5 - 6 | 679 | 62 |
BC112199.1 | - | 5 - 6 | 664 | 62 | |
BC143741.1 | - | 5 - 6 | 652 | 62 | |
BC143747.1 | - | 5 - 6 | 652 | 62 | |
BC143749.1 | - | 5 - 6 | 652 | 62 | |
L07868.1 | - | 5 - 6 | 658 | 62 |