The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018448.4 | NP_060918.2 | 4 - 5 | 931 | 93 |
GenBank mRNA | AB020636.2 | - | 4 - 5 | 833 | 93 |
AK027783.1 | - | 1 - 2 | 140 | 93 | |
AK074922.1 | - | 1 - 2 | 105 | 93 | |
AK096010.1 | - | 4 - 5 | 837 | 93 | |
AK292456.1 | - | 6 - 7 | 770 | 93 | |
AK314358.1 | - | 4 - 5 | 819 | 93 | |
AL136810.1 | - | 4 - 5 | 843 | 93 | |
BC026220.2 | - | 2 - 3 | 282 | 93 | |
BC050341.1 | - | 4 - 5 | 627 | 93 |