The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018072.1 | NP_001018082.1 | 2 - 3 | 1807 | 88 |
XM_005268645.3 | XP_005268702.1 | 2 - 3 | 175 | 88 | |
XM_011537909.2 | XP_011536211.1 | 2 - 3 | 730 | 88 | |
XM_011537910.2 | XP_011536212.1 | 2 - 3 | 684 | 88 | |
XM_017018807.1 | XP_016874296.1 | 2 - 3 | 3059 | 88 | |
GenBank mRNA | AK122818.1 | - | 2 - 3 | 306 | 88 |
AK124835.1 | - | 2 - 3 | 275 | 88 | |
BC146824.1 | - | 2 - 3 | 1806 | 88 |