The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016433.3 | NP_057517.1 | 4 - 5 | 564 | 73 |
XM_011538434.2 | XP_011536736.1 | 5 - 6 | 856 | 73 | |
GenBank mRNA | AF209704.1 | - | 4 - 5 | 460 | 73 |
AK313457.1 | - | 4 - 5 | 453 | 73 | |
AY372530.1 | - | 4 - 5 | 450 | 73 | |
AY372531.1 | - | 4 - 5 | 450 | 73 | |
AY372532.1 | - | 4 - 5 | 479 | 73 | |
BC009932.2 | - | 4 - 5 | 564 | 73 | |
JN039379.1 | - | 5 - 6 | 789 | 73 | |
JN052207.1 | - | 2 - 3 | 257 | 73 |