Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIF16B
Entrez Gene ID
55614
Gene Name
kinesin family member 16B
Gene Aliases
C20orf23, KISC20ORF, SNX23
Chromosome Location
Chr.20: 16272104 - 16573457 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199866.1 NP_001186795.1 22 - 23 3660 82
GenBank mRNA AB046810.2 - 22 - 23 3577 82
BC150261.1 - 22 - 23 3577 82

View Full Product Details