The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135993.1 | NP_001129465.1 | 9 - 10 | 1749 | 149 |
NM_001292030.1 | NP_001278959.1 | 4 - 5 | 626 | 149 | |
NM_153211.3 | NP_694943.2 | 9 - 10 | 1396 | 149 | |
XM_005258198.4 | XP_005258255.1 | 11 - 12 | 1783 | 149 | |
XM_011525814.2 | XP_011524116.1 | 8 - 9 | 1829 | 149 | |
XM_017025550.1 | XP_016881039.1 | 11 - 12 | 1877 | 149 | |
GenBank mRNA | AK091080.1 | - | 9 - 10 | 1396 | 149 |
AK296442.1 | - | 4 - 5 | 626 | 149 | |
BC032684.1 | - | 9 - 10 | 1198 | 149 | |
BC121034.2 | - | 9 - 10 | 1279 | 149 | |
BC121035.2 | - | 9 - 10 | 1279 | 149 |