The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006833.4 | NP_006824.2 | 3 - 4 | 359 | 92 |
GenBank mRNA | AK312024.1 | - | 3 - 4 | 350 | 92 |
BC002520.2 | - | 3 - 4 | 333 | 92 | |
CB990467.1 | - | 3 - 4 | 383 | 92 | |
U70735.1 | - | 3 - 4 | 281 | 92 |