The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006833.4 | NP_006824.2 | 8 - 9 | 771 | 78 |
GenBank mRNA | AK312024.1 | - | 8 - 9 | 762 | 78 |
BC002520.2 | - | 8 - 9 | 745 | 78 | |
U70735.1 | - | 8 - 9 | 693 | 78 |