The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001169117.1 | NP_001162588.1 | 7 - 8 | 1508 | 93 |
NM_001169118.1 | NP_001162589.1 | 7 - 8 | 1508 | 93 | |
NM_020860.3 | NP_065911.3 | 7 - 8 | 1508 | 93 | |
GenBank mRNA | AB040915.1 | - | 7 - 8 | 1196 | 93 |
AF328905.1 | - | 7 - 8 | 1510 | 93 | |
AK023369.1 | - | 1 - 2 | 104 | 93 | |
AK096846.1 | - | 7 - 8 | 1359 | 93 | |
AL136577.1 | - | 7 - 8 | 1051 | 93 | |
BC015659.2 | - | 3 - 4 | 407 | 93 | |
BC057231.1 | - | 1 - 2 | 61 | 93 | |
BC136449.1 | - | 7 - 8 | 1307 | 93 | |
BC146661.1 | - | 7 - 8 | 1404 | 93 | |
BC152554.1 | - | 7 - 8 | 1404 | 93 | |
BC171766.1 | - | 7 - 8 | 1306 | 93 |