The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286609.1 | NP_001273538.1 | 15 - 16 | 2391 | 69 |
NM_032131.5 | NP_115507.4 | 16 - 17 | 2464 | 69 | |
XM_005267154.3 | XP_005267211.1 | 16 - 17 | 2745 | 69 | |
XM_005267155.2 | XP_005267212.1 | 16 - 17 | 2480 | 69 | |
XM_005267157.3 | XP_005267214.1 | 16 - 17 | 2388 | 69 | |
XM_006715573.2 | XP_006715636.1 | 17 - 18 | 2570 | 69 | |
XM_006715574.2 | XP_006715637.1 | 13 - 14 | 1900 | 69 | |
XM_011536166.1 | XP_011534468.1 | 17 - 18 | 2555 | 69 | |
XM_011536167.1 | XP_011534469.1 | 16 - 17 | 2341 | 69 | |
XM_011536168.2 | XP_011534470.1 | 16 - 17 | 2458 | 69 | |
XM_011536172.2 | XP_011534474.1 | 15 - 16 | 2315 | 69 | |
XM_017011350.1 | XP_016866839.1 | 15 - 16 | 2393 | 69 | |
XM_017011352.1 | XP_016866841.1 | 15 - 16 | 2294 | 69 | |
GenBank mRNA | AK292499.1 | - | 16 - 17 | 2458 | 69 |
AK294574.1 | - | 15 - 16 | 2391 | 69 | |
AL136754.1 | - | 12 - 13 | 1814 | 69 | |
BC030603.2 | - | 16 - 17 | 2458 | 69 |