Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FLI1
Entrez Gene ID
2313
Gene Name
Fli-1 proto-oncogene, ETS transcription factor
Gene Aliases
EWSR2, SIC-1
Chromosome Location
Chr.11: 128685263 - 128813267 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017017406.1 XP_016872895.1 1 - 2 304 118

View Full Product Details