The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005103.4 | NP_005094.1 | 7 - 8 | 1253 | 67 |
XM_005271734.2 | XP_005271791.1 | 7 - 8 | 1298 | 67 | |
XM_005271735.2 | XP_005271792.1 | 7 - 8 | 1350 | 67 | |
GenBank mRNA | BC009545.1 | - | 7 - 8 | 1170 | 67 |
CR456817.1 | - | 6 - 7 | 1018 | 67 | |
CR541954.1 | - | 6 - 7 | 1018 | 67 | |
U60060.1 | - | 7 - 8 | 1117 | 67 | |
U69139.1 | - | 6 - 7 | 1043 | 67 |