Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLMP
Entrez Gene ID
79827
Gene Name
CXADR like membrane protein
Gene Aliases
ACAM, ASAM, CSBM, CSBS
Chromosome Location
Chr.11: 123072006 - 123195305 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024769.3 NP_079045.1 4 - 5 928 103
XM_011542998.2 XP_011541300.1 4 - 5 896 103
GenBank mRNA AK026068.1 - 4 - 5 904 103
AY326422.1 - 4 - 5 752 103
AY358340.1 - 4 - 5 752 103
BC009371.2 - 4 - 5 574 103

View Full Product Details