Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DIXDC1
Entrez Gene ID
85458
Gene Name
DIX domain containing 1
Gene Aliases
CCD1
Chromosome Location
Chr.11: 111927144 - 112022650 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037954.3 NP_001033043.1 8 - 9 1310 83
NM_033425.4 NP_219493.1 4 - 5 843 83
XM_017018466.1 XP_016873955.1 9 - 10 1270 83
XM_017018467.1 XP_016873956.1 9 - 10 1138 83
XM_017018468.1 XP_016873957.1 4 - 5 454 83
XM_017018469.1 XP_016873958.1 5 - 6 479 83
GenBank mRNA AB051522.1 - 4 - 5 750 83
AK055899.1 - 2 - 3 366 83
AK289807.1 - 4 - 5 819 83
BC033034.2 - 4 - 5 802 83
DQ642016.1 - 8 - 9 1310 83

View Full Product Details