Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DIXDC1
Entrez Gene ID
85458
Gene Name
DIX domain containing 1
Gene Aliases
CCD1
Chromosome Location
Chr.11: 111927144 - 112022650 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037954.3 NP_001033043.1 11 - 12 1520 63
NM_033425.4 NP_219493.1 7 - 8 1053 63
XM_017018466.1 XP_016873955.1 12 - 13 1480 63
XM_017018467.1 XP_016873956.1 12 - 13 1348 63
XM_017018468.1 XP_016873957.1 7 - 8 664 63
XM_017018469.1 XP_016873958.1 8 - 9 689 63
GenBank mRNA AB051522.1 - 7 - 8 960 63
AK055899.1 - 5 - 6 576 63
AK289807.1 - 7 - 8 1029 63
AK294954.1 - 2 - 3 219 63
BC033034.2 - 7 - 8 1012 63
BC041626.1 - 4 - 5 247 63
DQ642016.1 - 11 - 12 1520 63

View Full Product Details