The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025003.3 | NP_079279.3 | 38 - 39 | 5637 | 104 |
XM_011538754.2 | XP_011537056.1 | 38 - 39 | 5690 | 104 | |
XM_017019979.1 | XP_016875468.1 | 31 - 32 | 4706 | 104 | |
GenBank mRNA | AF488804.1 | - | 38 - 39 | 5640 | 104 |