Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC12A4
Entrez Gene ID
6560
Gene Name
solute carrier family 12 member 4
Gene Aliases
CTC-479C5.17, KCC1, hKCC1
Chromosome Location
Chr.16: 67943474 - 67968694 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK097808.1 - 10 - 11 1399 65

View Full Product Details