Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCAF8
Entrez Gene ID
22828
Gene Name
SR-related CTD associated factor 8
Gene Aliases
RBM16
Chromosome Location
Chr.6: 154733378 - 154834244 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286188.1 NP_001273117.1 2 - 3 313 70
NM_001286194.1 NP_001273123.1 2 - 3 214 70
NM_001286199.1 NP_001273128.1 2 - 3 266 70
NM_014892.4 NP_055707.3 1 - 2 557 70
GenBank mRNA AB029039.1 - 1 - 2 219 70
AK295651.1 - 2 - 3 275 70
AK302969.1 - 2 - 3 199 70
BC032728.1 - 1 - 2 519 70
BC070071.1 - 2 - 3 266 70
DA219101.1 - 1 - 2 520 70

View Full Product Details