The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306177.1 | NP_001293106.1 | 5 - 6 | 737 | 153 |
NM_004840.2 | NP_004831.1 | 6 - 7 | 1199 | 153 | |
XM_005262499.2 | XP_005262556.1 | 6 - 7 | 1511 | 153 | |
XM_011531412.2 | XP_011529714.1 | 7 - 8 | 1592 | 153 | |
XM_011531413.1 | XP_011529715.1 | 6 - 7 | 1511 | 153 | |
XM_011531414.1 | XP_011529716.1 | 6 - 7 | 1511 | 153 | |
XM_011531415.2 | XP_011529717.1 | 6 - 7 | 947 | 153 | |
XM_011531416.2 | XP_011529718.1 | 6 - 7 | 1210 | 153 | |
XM_011531417.1 | XP_011529719.1 | 5 - 6 | 1129 | 153 | |
XM_017029975.1 | XP_016885464.1 | 7 - 8 | 1592 | 153 | |
XM_017029976.1 | XP_016885465.1 | 5 - 6 | 1129 | 153 | |
GenBank mRNA | AK291742.1 | - | 6 - 7 | 742 | 153 |
AK294929.1 | - | 5 - 6 | 543 | 153 | |
AK295709.1 | - | 6 - 7 | 819 | 153 | |
AK299861.1 | - | 5 - 6 | 845 | 153 | |
BC039856.1 | - | 6 - 7 | 1199 | 153 | |
BC043505.1 | - | 5 - 6 | 703 | 153 | |
BX537390.1 | - | 6 - 7 | 804 | 153 | |
D25304.1 | - | 6 - 7 | 730 | 153 |