Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB16
Entrez Gene ID
7704
Gene Name
zinc finger and BTB domain containing 16
Gene Aliases
PLZF, ZNF145
Chromosome Location
Chr.11: 114059576 - 114256765 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC026902.1 - 1 - 2 76 73

View Full Product Details