The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175058.4 | NP_778228.3 | 6 - 7 | 540 | 87 |
XM_017017240.1 | XP_016872729.1 | 6 - 7 | 547 | 87 | |
XM_017017241.1 | XP_016872730.1 | 6 - 7 | 547 | 87 | |
XM_017017242.1 | XP_016872731.1 | 6 - 7 | 547 | 87 | |
XM_017017243.1 | XP_016872732.1 | 6 - 7 | 547 | 87 | |
GenBank mRNA | BC071599.1 | - | 6 - 7 | 536 | 87 |