The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012448.3 | NP_036580.2 | 16 - 17 | 2248 | 98 |
XM_017024977.1 | XP_016880466.1 | 15 - 16 | 2067 | 98 | |
GenBank mRNA | AB208920.1 | - | 16 - 17 | 2227 | 98 |
AK308163.1 | - | 16 - 17 | 2136 | 98 | |
BC065227.1 | - | 16 - 17 | 2248 | 98 | |
U47686.1 | - | 16 - 17 | 2225 | 98 | |
U48730.2 | - | 15 - 16 | 2090 | 98 |