The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182503.2 | NP_872309.2 | 1 - 2 | 146 | 76 |
GenBank mRNA | BC037955.1 | - | 1 - 2 | 106 | 76 |