The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286259.1 | NP_001273188.1 | 4 - 5 | 1605 | 68 |
NM_182503.2 | NP_872309.2 | 4 - 5 | 507 | 68 | |
XM_011535439.2 | XP_011533741.1 | 7 - 8 | 663 | 68 | |
XM_011535440.1 | XP_011533742.1 | 7 - 8 | 3395 | 68 | |
XM_011535444.2 | XP_011533746.1 | 6 - 7 | 526 | 68 | |
XM_017010260.1 | XP_016865749.1 | 5 - 6 | 3924 | 68 | |
XM_017010261.1 | XP_016865750.1 | 4 - 5 | 3881 | 68 | |
XM_017010262.1 | XP_016865751.1 | 4 - 5 | 385 | 68 | |
XM_017010263.1 | XP_016865752.1 | 3 - 4 | 359 | 68 | |
GenBank mRNA | AK126201.1 | - | 4 - 5 | 1611 | 68 |
BC037955.1 | - | 4 - 5 | 467 | 68 | |
BX538182.1 | - | 4 - 5 | 905 | 68 |