The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282166.1 | NP_001269095.1 | 3 - 4 | 1123 | 89 |
NM_003173.3 | NP_003164.1 | 3 - 4 | 876 | 89 | |
GenBank mRNA | AF019968.1 | - | 3 - 4 | 869 | 89 |
AK223071.1 | - | 3 - 4 | 848 | 89 | |
AK298233.1 | - | 4 - 5 | 582 | 89 | |
AK299900.1 | - | 3 - 4 | 1123 | 89 | |
AK312547.1 | - | 3 - 4 | 848 | 89 | |
BC006238.2 | - | 3 - 4 | 867 | 89 | |
CR541746.1 | - | 3 - 4 | 824 | 89 | |
L08238.1 | - | 2 - 3 | 964 | 90 |