Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KDM2A
Entrez Gene ID
22992
Gene Name
lysine demethylase 2A
Gene Aliases
CXXC8, FBL11, FBL7, FBXL11, JHDM1A, LILINA
Chromosome Location
Chr.11: 67119210 - 67258082 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012308.2 NP_036440.1 9 - 10 1706 145
NR_027473.1 9 - 10 1706 145
XM_006718480.3 XP_006718543.1 5 - 6 724 145
XM_011544860.2 XP_011543162.1 9 - 10 1765 145
XM_011544861.1 XP_011543163.1 4 - 5 418 145
XM_017017391.1 XP_016872880.1 9 - 10 1012 145
XM_017017392.1 XP_016872881.1 8 - 9 823 145
GenBank mRNA AB023221.2 - 9 - 10 1023 145
AK024505.1 - 9 - 10 948 145
BC047371.1 - 4 - 5 418 145
BC047486.1 - 9 - 10 1708 145
BC064360.1 - 9 - 10 1121 145
JQ710741.1 - 8 - 9 890 145
JQ710742.1 - 9 - 10 1012 145
JQ710743.1 - 8 - 9 890 145
JQ710744.1 - 10 - 11 1059 145

View Full Product Details