Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CXorf21
Entrez Gene ID
80231
Gene Name
chromosome X open reading frame 21
Gene Aliases
-
Chromosome Location
Chr.X: 30558824 - 30577916 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025159.2 NP_079435.1 2 - 3 401 147
GenBank mRNA AK021639.1 - 2 - 3 282 147
BC020611.1 - 2 - 3 329 147
DA921873.1 - 2 - 3 401 147

View Full Product Details