The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000440.2 | NP_000431.2 | 15 - 16 | 2042 | 74 |
XM_011537650.2 | XP_011535952.1 | 12 - 13 | 1173 | 74 | |
XM_011537651.2 | XP_011535953.1 | 9 - 10 | 1641 | 74 | |
XM_011537652.1 | XP_011535954.1 | 10 - 11 | 1111 | 74 | |
XM_011537653.2 | XP_011535955.1 | 9 - 10 | 1807 | 74 | |
XM_011537654.2 | XP_011535956.1 | 9 - 10 | 1804 | 74 | |
XM_017009572.1 | XP_016865061.1 | 13 - 14 | 1799 | 74 | |
GenBank mRNA | AB593141.1 | - | 13 - 14 | 1799 | 74 |
BC035909.1 | - | 15 - 16 | 2029 | 74 | |
BC144044.1 | - | 13 - 14 | 2033 | 74 | |
M26061.1 | - | 15 - 16 | 2042 | 74 |