The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000440.2 | NP_000431.2 | 20 - 21 | 2479 | 97 |
XM_011537650.2 | XP_011535952.1 | 17 - 18 | 1610 | 97 | |
XM_011537651.2 | XP_011535953.1 | 14 - 15 | 2078 | 97 | |
XM_011537652.1 | XP_011535954.1 | 15 - 16 | 1548 | 97 | |
XM_011537653.2 | XP_011535955.1 | 14 - 15 | 2244 | 97 | |
XM_011537654.2 | XP_011535956.1 | 14 - 15 | 2241 | 97 | |
XM_017009572.1 | XP_016865061.1 | 18 - 19 | 2236 | 97 | |
GenBank mRNA | AB593141.1 | - | 18 - 19 | 2236 | 97 |
BC035909.1 | - | 20 - 21 | 2466 | 97 | |
BC144044.1 | - | 18 - 19 | 2470 | 97 | |
BX648986.1 | - | 1 - 2 | 1724 | 97 | |
M26061.1 | - | 20 - 21 | 2479 | 97 |