Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSD2
Entrez Gene ID
84249
Gene Name
pleckstrin and Sec7 domain containing 2
Gene Aliases
EFA6C
Chromosome Location
Chr.5: 139683980 - 139844466 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032289.2 NP_115665.1 10 - 11 1796 77
XM_011537696.2 XP_011535998.1 11 - 12 1852 77
XM_017009975.1 XP_016865464.1 11 - 12 3159 77
XM_017009976.1 XP_016865465.1 11 - 12 3967 77
XM_017009977.1 XP_016865466.1 11 - 12 3847 77
GenBank mRNA AL136559.1 - 10 - 11 1796 77
AL834268.1 - 6 - 7 578 77
BC038233.1 - 10 - 11 1773 77

View Full Product Details