The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032289.2 | NP_115665.1 | 4 - 5 | 1218 | 63 |
XM_011537696.2 | XP_011535998.1 | 5 - 6 | 1274 | 63 | |
XM_017009975.1 | XP_016865464.1 | 5 - 6 | 2581 | 63 | |
XM_017009976.1 | XP_016865465.1 | 5 - 6 | 3389 | 63 | |
XM_017009977.1 | XP_016865466.1 | 5 - 6 | 3269 | 63 | |
GenBank mRNA | AL136559.1 | - | 4 - 5 | 1218 | 63 |
BC038233.1 | - | 4 - 5 | 1195 | 63 |