The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145933.1 | NP_001139405.1 | 4 - 5 | 709 | 70 |
NM_001145934.1 | NP_001139406.1 | 3 - 4 | 654 | 70 | |
NM_012253.3 | NP_036385.3 | 4 - 5 | 727 | 70 | |
GenBank mRNA | AK292224.1 | - | 4 - 5 | 599 | 70 |
AK292261.1 | - | 4 - 5 | 603 | 70 | |
AK301724.1 | - | 4 - 5 | 528 | 70 | |
AK302044.1 | - | 4 - 5 | 585 | 70 | |
BC025382.2 | - | 4 - 5 | 603 | 70 | |
X91817.1 | - | 3 - 4 | 493 | 70 |