Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SUV39H1
Entrez Gene ID
6839
Gene Name
suppressor of variegation 3-9 homolog 1
Gene Aliases
H3-K9-HMTase 1, KMT1A, MG44, SUV39H
Chromosome Location
Chr.X: 48695554 - 48709016 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282166.1 NP_001269095.1 1 - 2 317 79
GenBank mRNA AK299900.1 - 1 - 2 317 79
L08238.1 - - 79

View Full Product Details