The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144495.2 | NP_652766.1 | 4 - 5 | 478 | 87 |
XM_005272572.4 | XP_005272629.1 | 4 - 5 | 401 | 87 | |
GenBank mRNA | AB041836.1 | - | 3 - 4 | 289 | 87 |
AJ973593.1 | - | 3 - 4 | 304 | 87 | |
AJ973607.1 | - | 3 - 4 | 287 | 87 |