Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BEST1
Entrez Gene ID
7439
Gene Name
bestrophin 1
Gene Aliases
ARB, BEST, BMD, RP50, TU15B, VMD2
Chromosome Location
Chr.11: 61946722 - 61964465 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300786.1 NP_001287715.1 8 - 9 2054 101
NM_001300787.1 NP_001287716.1 9 - 10 2135 101
NM_004183.3 NP_004174.1 10 - 11 2323 101
NR_134580.1 10 - 11 2526 101
XM_005274215.3 XP_005274272.1 8 - 9 2088 101
GenBank mRNA AB209451.1 - 6 - 7 5575 101
AF057169.1 - 10 - 11 1847 101
AF057170.1 - 10 - 11 2050 101
AF073501.1 - 10 - 11 1831 101
AK295998.1 - 9 - 10 1669 101
AK301392.1 - 8 - 9 1652 101
BC015220.2 - 8 - 9 2054 101
JQ954696.1 - 9 - 10 1809 101

View Full Product Details