The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001549.2 | NP_001001549.1 | 7 - 8 | 879 | 95 |
GenBank mRNA | U34355.1 | - | 7 - 8 | 879 | 95 |