Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALG8
Entrez Gene ID
79053
Gene Name
ALG8, alpha-1,3-glucosyltransferase
Gene Aliases
CDG1H
Chromosome Location
Chr.11: 78100942 - 78139660 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007027.2 NP_001007028.1 2 - 3 236 110
NM_024079.4 NP_076984.2 2 - 3 236 110
XM_005274247.3 XP_005274304.1 2 - 3 997 110
XM_011545252.2 XP_011543554.1 2 - 3 246 110
XM_017018274.1 XP_016873763.1 2 - 3 984 110
GenBank mRNA AJ224875.1 - 2 - 3 190 110
AK075184.1 - 2 - 3 175 111
AK075194.1 - 2 - 3 191 110
AK098614.1 - 2 - 3 236 110
AK309050.1 - 2 - 3 205 110
BC001133.2 - 2 - 3 219 110
KU178764.1 - 2 - 3 171 110

View Full Product Details