The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002711.3 | NP_002702.2 | 1 - 2 | 808 | 152 |
XM_005250473.3 | XP_005250530.1 | 2 - 3 | 354 | 152 | |
GenBank mRNA | AF024579.1 | - | 2 - 3 | 256 | 152 |
BC056666.1 | - | 1 - 2 | 811 | 152 | |
BC070313.1 | - | 1 - 2 | 790 | 152 | |
BC126451.1 | - | 1 - 2 | 846 | 152 | |
BC126453.1 | - | 1 - 2 | 846 | 152 | |
X78578.1 | - | 1 - 2 | 775 | 152 |