The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000440.2 | NP_000431.2 | 2 - 3 | 740 | 115 |
GenBank mRNA | BC035909.1 | - | 2 - 3 | 727 | 115 |
BC144044.1 | - | 2 - 3 | 667 | 115 |