The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002972.3 | NP_002963.2 | 3 - 4 | 512 | 76 |
XM_005261935.2 | XP_005261992.1 | 3 - 4 | 460 | 76 | |
XM_011530710.2 | XP_011529012.1 | 3 - 4 | 326 | 76 | |
GenBank mRNA | BC040031.1 | - | 3 - 4 | 463 | 76 |
BC087612.1 | - | 3 - 4 | 451 | 76 | |
BU931657.1 | - | - | 474 | 76 |