Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLAUR
Entrez Gene ID
5329
Gene Name
plasminogen activator, urokinase receptor
Gene Aliases
CD87, U-PAR, UPAR, URKR
Chromosome Location
Chr.19: 43646095 - 43670346 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005377.2 NP_001005377.1 5 - 6 844 76
NM_002659.3 NP_002650.1 6 - 7 979 76
XM_011527027.2 XP_011525329.1 5 - 6 657 76
XM_011527028.2 XP_011525330.1 6 - 7 821 76
XM_017026872.1 XP_016882361.1 6 - 7 1214 76
XM_017026873.1 XP_016882362.1 4 - 5 867 76
GenBank mRNA A18757.1 - 6 - 7 799 76
AK290774.1 - 6 - 7 799 76
AY029180.1 - 6 - 7 799 76
BC002788.2 - 6 - 7 818 76
CD370232.1 - 1 - 2 76
M83246.1 - 6 - 7 770 76
U08839.1 - 5 - 6 1044 76
X51675.1 - 6 - 7 799 76

View Full Product Details