Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLAUR
Entrez Gene ID
5329
Gene Name
plasminogen activator, urokinase receptor
Gene Aliases
CD87, U-PAR, UPAR, URKR
Chromosome Location
Chr.19: 43646095 - 43670346 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005376.2 NP_001005376.1 5 - 6 835 69
NM_002659.3 NP_002650.1 5 - 6 835 69
XM_005258990.4 XP_005259047.1 5 - 6 657 69
XM_011527027.2 XP_011525329.1 4 - 5 513 69
XM_017026872.1 XP_016882361.1 5 - 6 1070 69
XM_017026873.1 XP_016882362.1 3 - 4 723 69
GenBank mRNA A18757.1 - 5 - 6 655 69
AF257789.1 - 5 - 6 609 69
AK290774.1 - 5 - 6 655 69
AY029180.1 - 5 - 6 655 69
BC002788.2 - 5 - 6 674 69
CR456952.1 - 5 - 6 609 69
M83246.1 - 5 - 6 626 69
X51675.1 - 5 - 6 655 69
X74039.1 - 5 - 6 655 69

View Full Product Details