Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
XRCC1
Entrez Gene ID
7515
Gene Name
X-ray repair cross complementing 1
Gene Aliases
RCC
Chromosome Location
Chr.19: 43543312 - 43575578 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006297.2 NP_006288.2 5 - 6 613 96
GenBank mRNA AB208781.1 - 5 - 6 536 96
AK293542.1 - 5 - 6 591 96
AK315332.1 - 5 - 6 602 96
BC023593.2 - 5 - 6 500 96
CR456728.1 - 5 - 6 493 96
M36089.1 - 5 - 6 1041 96

View Full Product Details