Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SYT7
Entrez Gene ID
9066
Gene Name
synaptotagmin 7
Gene Aliases
IPCA-7, IPCA7, PCANAP7, SYT-VII, SYTVII
Chromosome Location
Chr.11: 61513716 - 61588404 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252065.1 NP_001238994.1 10 - 10 83
NM_001300773.1 NP_001287702.1 10 - 10 83
NM_004200.3 NP_004191.2 9 - 9 83
GenBank mRNA BM675410.1 - 1 - 1 186 83
BQ447018.1 - 1 - 1 189 83

View Full Product Details