The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020369.2 | NP_065102.1 | 5 - 6 | 1512 | 90 |
GenBank mRNA | AF281049.1 | - | 5 - 6 | 1512 | 90 |
AK314050.1 | - | 5 - 6 | 1417 | 90 | |
BC035606.1 | - | 5 - 6 | 1387 | 90 |