The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014207.3 | NP_055022.2 | 8 - 9 | 1377 | 130 |
GenBank mRNA | AK292698.1 | - | 8 - 9 | 1346 | 130 |
BC027901.1 | - | 8 - 9 | 1340 | 130 | |
X04391.1 | - | 8 - 9 | 1346 | 130 |