The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308103.1 | NP_001295032.1 | 2 - 3 | 267 | 106 |
NM_022073.3 | NP_071356.1 | 2 - 3 | 801 | 106 | |
GenBank mRNA | AJ310545.1 | - | 2 - 3 | 801 | 106 |
AK025273.1 | - | 2 - 3 | 801 | 106 | |
AK123350.1 | - | 3 - 4 | 630 | 106 | |
AK225473.1 | - | 2 - 3 | 801 | 106 | |
BC010992.2 | - | 2 - 3 | 757 | 106 | |
BC064924.1 | - | 2 - 3 | 761 | 106 | |
BC102030.1 | - | 2 - 3 | 267 | 106 | |
BC105938.1 | - | 2 - 3 | 267 | 106 | |
BC105939.1 | - | 2 - 3 | 549 | 106 | |
BC111057.1 | - | 2 - 3 | 784 | 106 | |
BX346941.2 | - | 1 - 2 | 124 | 106 | |
DQ975379.1 | - | 2 - 3 | 201 | 106 |