Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRL
Entrez Gene ID
6345
Gene Name
sarcalumenin
Gene Aliases
-
Chromosome Location
Chr.16: 4189374 - 4242253 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098814.1 NP_001092284.1 5 - 6 628 72
NM_001323667.1 NP_001310596.1 6 - 7 758 72
NM_001323668.1 NP_001310597.1 6 - 7 2006 72
XM_017023527.1 XP_016879016.1 6 - 7 1953 72
XM_017023528.1 XP_016879017.1 5 - 6 659 72
XM_017023529.1 XP_016879018.1 5 - 6 674 72
GenBank mRNA AK056588.1 - 5 - 6 628 72
AK302601.1 - 5 - 6 664 72
AL831973.2 - 5 - 6 600 72
AL832361.1 - 6 - 7 2000 72
BX647476.1 - 6 - 7 2000 72

View Full Product Details