The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042729.1 | NP_001036194.1 | 2 - 3 | 200 | 60 |
NM_001042747.1 | NP_001036212.1 | 2 - 3 | 259 | 60 | |
NM_005248.2 | NP_005239.1 | 2 - 3 | 216 | 60 | |
XM_006710452.2 | XP_006710515.1 | 3 - 4 | 267 | 60 | |
XM_011541010.1 | XP_011539312.1 | 2 - 3 | 181 | 60 | |
XM_011541011.2 | XP_011539313.1 | 2 - 3 | 187 | 60 | |
XM_011541012.1 | XP_011539314.1 | 2 - 3 | 216 | 60 | |
XM_011541013.2 | XP_011539315.1 | 2 - 3 | 216 | 60 | |
GenBank mRNA | AK125143.1 | - | 2 - 3 | 103 | 60 |
AK130188.1 | - | 2 - 3 | 178 | 60 | |
AK301078.1 | - | 2 - 3 | 172 | 60 | |
AK304846.1 | - | 2 - 3 | 199 | 60 | |
BC064382.1 | - | 2 - 3 | 129 | 60 | |
BI771758.1 | - | 2 - 3 | 259 | 60 | |
BX495937.1 | - | 2 - 3 | 260 | 60 | |
DA139476.1 | - | 2 - 3 | 216 | 60 | |
DA952771.1 | - | 2 - 3 | 155 | 60 | |
DA960178.1 | - | 2 - 3 | 183 | 60 | |
M27454.1 | - | 2 - 3 | 141 | 60 |