The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042729.1 | NP_001036194.1 | 7 - 8 | 888 | 92 |
NM_001042747.1 | NP_001036212.1 | 7 - 8 | 947 | 92 | |
NM_005248.2 | NP_005239.1 | 7 - 8 | 904 | 92 | |
XM_006710452.2 | XP_006710515.1 | 8 - 9 | 955 | 92 | |
XM_011541010.1 | XP_011539312.1 | 7 - 8 | 869 | 92 | |
XM_011541011.2 | XP_011539313.1 | 5 - 6 | 668 | 92 | |
XM_011541012.1 | XP_011539314.1 | 7 - 8 | 904 | 92 | |
XM_011541013.2 | XP_011539315.1 | 7 - 8 | 904 | 92 | |
XM_011541014.2 | XP_011539316.1 | 3 - 4 | 486 | 92 | |
XM_017000673.1 | XP_016856162.1 | 5 - 6 | 524 | 92 | |
XM_017000674.1 | XP_016856163.1 | 4 - 5 | 459 | 92 | |
GenBank mRNA | AK130188.1 | - | 7 - 8 | 866 | 92 |
AK301078.1 | - | 5 - 6 | 662 | 92 | |
BC064382.1 | - | 7 - 8 | 817 | 92 | |
M19722.1 | - | 5 - 6 | 823 | 92 |